Learn More
Agilent BioTek ELx405 Select Microplate Washer, IVD
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek ELX405CVSI
Description
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.
Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
| CE and TUV marked. RoHS compliant. | |
| ≤2 μL/well | |
| Microplate Washer | |
| 1 to 10 | |
| One positive displacement pump | |
| 50/60 Hz | |
| 59°F to 86°F | |
| 96-well manifold; Automatic 4-Buffer Switching | |
| 35.6 cm | |
| 25.4 cm | |
| 43.2 cm | |
| 100/240 V |
| ≤3% CV | |
| Programmable in minutes and seconds, up to 60 min. | |
| 13.5 kg | |
| Backlit LCD | |
| 10 to 80%, noncondensing | |
| ELX405CV | |
| 15°C to 30°C | |
| 14 in. | |
| 10 in. | |
| 17 in. | |
| 30 lb. | |
| <8 A |