missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek ELx405 Select Microplate Washer, IVD
GREENER_CHOICE

Catalog No. BTELX405SI
Change view
Click to view available options
Model:
ELX405
ELX405CS
ELX405CV
ELX405CVS
Includes:
96-well manifold
96-well manifold; Automatic 4-Buffer Switching
96-well manifold; Ultrasonic Advantage
96-well manifold; Ultrasonic Advantage; Automatic 4-Buffer Switching
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTELX405SI ELX405 96-well manifold
BTELX405CSI ELX405CS 96-well manifold; Ultrasonic Advantage
BTELX405CVI ELX405CV 96-well manifold; Automatic 4-Buffer Switching
BTELX405VSI ELX405CVS 96-well manifold; Ultrasonic Advantage; Automatic 4-Buffer Switching
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTELX405SI Supplier Agilent BioTek Supplier No. ELX405SI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. For In Vitro Diagnostic Use.

The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.

Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Precision ≤3% CV
Residual Volume ≤2 μL/well
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer
Weight (Metric) 13.5 kg
Wash Cycles 1 to 10
Display Backlit LCD
Fluid Delivery Type One positive displacement pump
Humidity 10 to 80%, noncondensing
Frequency 50/60 Hz
Model ELX405
Temperature (English) Operating 59°F to 86°F
Temperature (Metric) Operating 15°C to 30°C
Includes 96-well manifold
Width (English) 14 in.
Width (Metric) 35.6 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Depth (English) 17 in.
Depth (Metric) 43.2 cm
Weight (English) 30 lb.
Voltage 100/240 V
Amperage <8 A
Show More Show Less