missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek ELx405 Select Deep Well Microplate Washer

Description
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.
Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Certifications
CE and ETL marked
Specifications
Specifications
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Precision | ≤3% CV |
| Plate Type | 96 and 384-well Plates |
| Residual Volume | ≤2 μL/well |
| Soak Time | Programmable in minutes and seconds, up to 60 min. |
| Type | Microplate Washer |
| Weight (Metric) | 13.5 kg |
| Wash Cycles | 1 to 10 |
| Display | Backlit LCD |
| Fluid Delivery Type | One positive displacement pump |
| Show More |