missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eluent Solution, for Ion Chromatography, 1.7 mM NaHCO3 - 1.8 mM Na2CO3, Ricca Chemical
For Ion Chromatography
Specifications
| Chemical Name or Material | Eluent Solution |
|---|---|
| Name Note | 1.7mM NaHCO |
| CAS Min % | 98.0, 0.02, 0.01 |
| CAS Max % | 100.0, 0.02, 0.01 |
| CAS | 497-19-8 , 144-55-8 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
28101
|
N/A | 4 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
281032
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||