missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELP4 Recombinant Rabbit Monoclonal Antibody (23GB5310)
GREENER_CHOICE

Catalog No. MA553882
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553882 100 μL
MA553883 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553882 Supplier Invitrogen™ Supplier No. MA553882
Only null left

Rabbit Recombinant Monoclonal Antibody

ELP4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.

Specifications

Antigen ELP4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5310
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Elp4
Gene Accession No. Q96EB1, Q9ER73
Gene Alias A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog
Gene Symbols Elp4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ELP4 (313-364AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26610, 687694, 77766
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less