missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELOVL5 Recombinant Rabbit Monoclonal Antibody (23GB3580)
GREENER_CHOICE

Catalog No. MA552618
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552618 100 μL
MA552619 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552618 Supplier Invitrogen™ Supplier No. MA552618
Only null left

Rabbit Recombinant Monoclonal Antibody

ELOVL5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

ELOVL5 plays a role in elongation of long-chain polyunsaturated fatty acids.

Specifications

Antigen ELOVL5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3580
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ELOVL5
Gene Accession No. Q9NYP7
Gene Alias 1110059L23Rik; 3-keto acyl-CoA synthase ELOVL5; AI747313; AU043003; dJ483K16.1; elongation of long chain fatty acids family member 5; elongation of very long chain fatty acids protein 5; elongation of very long chain fatty acids-like 5; ELOVL FA elongase 5; ELOVL family member 5, elongation of long chain fatty acids; ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL family member 5, elongation of long chain fatty acids (yeast); ELOVL fatty acid elongase 5; ELOVL2; ELOVL5; Fatty acid elongase 1; hELO1; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2; PRO0530; rELO1; SCA38; spinocerebellar ataxia 38; very long chain 3-ketoacyl-CoA synthase 5; Very long chain 3-oxoacyl-CoA synthase 5; very-long-chain 3-oxoacyl-CoA synthase 5
Gene Symbols ELOVL5
Host Species Rabbit
Immunogen A synthetic peptide of human ELOVL5 (250-299AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60481
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less