missing translation for 'onlineSavingsMsg'
Learn More

Eleutheroside E, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014130
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014130 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014130 Supplier Merck Emd Millipore Supplier No. 0819810MG
Only null left

Eleutheroside E is one of the major bioactive saponins of Eleutherococcus senticosus.

Featured Industries: Food and Beverages

Specifications

CAS 39432-56-9
Linear Formula C34H46O18
Synonym [(1 R,3 aR,4 S,6 AS)-Tetrahydro-1 H,3 H-furo[3,4-c]furan-1,4-diyl]bis(2,6-dimethoxy-4,1-phenylene) bis-beta-D-glucopyranoside
Type Analytical Standard
Quantity 10 mg
Formula Weight 742.72
Percent Purity ≥98% (HPLC)
Grade Analytical Standard