Learn More
Cole-Parmer™ elements™ Modular Magnetic Stirrer
Optimize your stirring needs and workspace with this completely customizable stirrer
Supplier:  Cole-Parmer™ 8400382
 View more versions of this product
Catalog No. 11-475-001
Only null left
Explore available promotions
Description
Description
  • Attach up to 10 stir plates based on need
  • Simple touch and slide operation
  • LCD display with real time parameters
  • Corrosion-resistant glass stir top
  • Waterproof USB attachments
Specifications
Specifications
100 to 1500 rpm
Glass
Digital
100/240 V
50/60 Hz
1
1 L/position
Round
3.75 in.
Stirring Unit
Product Suggestions
Product Suggestions