missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Elemene, ≥98% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 515-13-9 |
| Flash Point | 98°C |
| Physical Form | Neat |
| Linear Formula | C15H24 |
| MDL Number | MFCD00468041 |
| Synonym | beta-Elemene; (1 S,2 S,4 R)-(-)-2,4-Diisopropenyl-1-methyl-1-vinylcyclohexane; (1 S,2 S,4 R)-1-Ethenyl-1-methyl-2,4-bis(1-methylethenyl)cyclohexane |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 204.35 |
| Percent Purity | ≥98% (GC) |
| Show More |