missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Electrode Storage Solution | |
| 98.4,1,0.6 | |
| Liquid | |
| 4 L |
| Poly Bottle | |
| 877-24-7,7447-40-7 | |
| KHC8H4O4 | |
| 204.23 |
| Electrode Storage Solution | |
| 98.4,1,0.6 | |
| Liquid | |
| 4 L |
| Poly Bottle | |
| 877-24-7,7447-40-7 | |
| KHC8H4O4 | |
| 204.23 |