missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrochemical Standards, 0.5M DIPA-HCL-ISA, Mettler Toledo™
Description
The new Sodium Analyzer determines the sodium content in food products simply and quickly. Reduce sample preparation time and effort by using safe and cheap chemicals while specifically and exclusively measuring sodium ions.
Specifications
Specifications
| Chemical Name or Material | 0.5M DIPA-HCL-ISA |
| Packaging | Cardboard |
| CAS | 108-18-9, 819-79-4, 7732-18-5 |
| Molecular Formula | C6H16ClNO2 |
| Color | Colorless |
| pH | 11.4 |
| Boiling Point | Approx. 100°C |
| Solution Type | Calibration Solution |
| Industry Type | Verification |
| Quantity | 25 mL |