missing translation for 'onlineSavingsMsg'
Learn More

Electrochemical Standards, 0.5M DIPA-HCL-ISA, Mettler Toledo™

Catalog No. 01912567
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912567 25 mL
01912568 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01912567 Supplier METTLER TOLEDO Supplier No. 30064081

For use with Sodium analyzer

The new Sodium Analyzer determines the sodium content in food products simply and quickly. Reduce sample preparation time and effort by using safe and cheap chemicals while specifically and exclusively measuring sodium ions.

Specifications

Chemical Name or Material 0.5M DIPA-HCL-ISA
Packaging Cardboard
CAS 108-18-9, 819-79-4, 7732-18-5
Molecular Formula C6H16ClNO2
Color Colorless
pH 11.4
Boiling Point Approx. 100°C
Solution Type Calibration Solution
Industry Type Verification
Quantity 25 mL