missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELAVL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596475
Description
Immunogen sequence: METQLSNGPT CNNTANGPTT INNNCSSPVD SGNTEDSKTN LIVNYLPQNM TQEELKSLFG SIGEIESCKL VRDKITGQSL GYGFVNYIDP KDAEKAINTL; Positive Samples: SH-SY5Y, Mouse spinal cord.
The protein encoded by this gene is a neural-specific RNA-binding protein that is known to bind to several 3' UTRs, including its own and also that of FOS and ID. The encoded protein may recognize a GAAA motif in the RNA. Three transcript variants encoding two different isoforms have been found for this gene.Chemical Identifiers
| ELAVL2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ELAVL2 | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV like neuron-specific RNA binding protein 2; Elav2; ELAVL 2; ELAVL 4; ELAVL2; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen B; Hu antigen D; hu-antigen B; HUB; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB; zgc:91918 | |
| ELAVL2 | |
| Primary | |
| Affinity Chromatography |
| 0.88 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12926, Q60899, Q8CH84 | |
| 15569, 1993, 286973 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ELAVL2 (NP_0044232). | |
| Antibody |
Specifications
| ELAVL2 | |
| Polyclonal | |
| Unconjugated | |
| ELAVL2 | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV like neuron-specific RNA binding protein 2; Elav2; ELAVL 2; ELAVL 4; ELAVL2; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen B; Hu antigen D; hu-antigen B; HUB; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB; zgc:91918 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15569, 1993, 286973 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12926, Q60899, Q8CH84 | |
| ELAVL2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ELAVL2 (NP_0044232). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |