Learn More
Agilent BioTek EL406 Microplate Washer/Dispenser

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers.
Supplier: Agilent BioTek 406SNA0LBSN
Description
The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. The EL406 capabilities include washing and dispensing for standard assays, plus biomagnetic separation and vacuum filtration protocols.
The Dual-Action manifold optimizes washing for loosely adherent cell monolayers. The patented Ultrasonic Advantage prevents blockages in the manifold tubes before washing, to reduce assay failure. This modular plate washer dispenser offers flexibility with both syringe pump and peristaltic pump dispensers, along with plate washing, on a single platform.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Certifications
CE marked; ETL listed
Chemical Identifiers
| ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL | |
| Optional (1 or 2 L) | |
| 500 nL to 3000 μL/well | |
| ≤2 μL/well | |
| Programmable in minutes and seconds, up to 60 min. | |
| 14.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 1 μL | |
| 42 cm | |
| 46 cm | |
| 100/240 V |
| CE and TUV marked. RoHS compliant. | |
| Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL | |
| 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates | |
| 96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s | |
| Microplate Washer/Dispenser | |
| 1 to 250 | |
| 50/60 Hz | |
| 16.5 in. | |
| 18 in. | |
| 32 lb. |
Specifications
| ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL | |
| Optional (1 or 2 L) | |
| 500 nL to 3000 μL/well | |
| ≤2 μL/well | |
| Programmable in minutes and seconds, up to 60 min. | |
| 14.5 kg | |
| High to Low Flow; Optimized rates for cell assays | |
| 406SNA0LB | |
| 1536-well microplate washing and dispensing (large bore, steel tubes); 2-Reagent Dispense Module – 2 Syringe Pumps (non-autoclavable/(1) 16-tube (2 x 8) manifolds - except with P/N 406SNA0-LB/steel tubes) | |
| 42 cm | |
| 46 cm | |
| 100/240 V |
| CE and TUV marked. RoHS compliant. | |
| Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL | |
| 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates | |
| 96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s | |
| Microplate Washer/Dispenser | |
| 1 to 250 | |
| 50/60 Hz | |
| 1 μL | |
| 16.5 in. | |
| 18 in. | |
| 32 lb. |