missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek EL406 Microplate Washer/Dispenser
GREENER_CHOICE

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers.

Supplier:  Agilent BioTek 406PSUB2SBSN

 View more versions of this product

Catalog No. B406PSUB2SB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. The EL406 capabilities include washing and dispensing for standard assays, plus biomagnetic separation and vacuum filtration protocols.

The Dual-Action manifold optimizes washing for loosely adherent cell monolayers. The patented Ultrasonic Advantage prevents blockages in the manifold tubes before washing, to reduce assay failure. This modular plate washer dispenser offers flexibility with both syringe pump and peristaltic pump dispensers, along with plate washing, on a single platform.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Certifications

CE marked; ETL listed

Specifications

Chemical Identifiers

±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL
Optional (1 or 2 L)
500 nL to 3000 μL/well
≤2 μL/well
Programmable in minutes and seconds, up to 60 min.
14.5 kg
High to Low Flow; Optimized rates for cell assays
1 μL
42 cm
46 cm
100/240 V
CE and TUV marked. RoHS compliant.
Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL
96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates
96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s
Microplate Washer/Dispenser
1 to 250
50/60 Hz
16.5 in.
18 in.
32 lb.

Specifications

±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL
Optional (1 or 2 L)
500 nL to 3000 μL/well
≤2 μL/well
Programmable in minutes and seconds, up to 60 min.
14.5 kg
High to Low Flow; Optimized rates for cell assays
406PSUB2SB
Faster 384-well microplate washing (192-tube (16 x 12) manifold/steel tubes); 1536-well microplate washing and dispensing (small bore, sapphire jeweled tubes); 1-Reagent Dispense Module – 1 Peristaltic Pump (autoclavable, 8-tip (1 x 8) cassette/sapphire jeweled, steel or plastic tips); 2-Reagent Dispense Module – 2 Syringe Pumps (autoclavable/(2) 16-tube (1 x 16) manifolds/steel tubes); Automatic 4-Buffer Switching
42 cm
31.8 cm
46 cm
100/240 V
CE and TUV marked. RoHS compliant.
Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL
96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates
96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s
Microplate Washer/Dispenser
1 to 250
50/60 Hz
1 μL
16.5 in.
12.5 in.
18 in.
32 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions