missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EKLF Monoclonal Antibody (1B6A3)
GREENER_CHOICE

Catalog No. PIMA538552
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538552 Supplier Invitrogen™ Supplier No. MA538552
Only null left

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.

Specifications

Antigen EKLF
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B6A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Klf1
Gene Accession No. Q13351
Gene Alias CDAN4; Eklf; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1
Gene Symbols Klf1
Host Species Mouse
Immunogen Purified recombinant fragment of human KLF1 (aa: 208-362) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less