missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3A/eIF3S10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302003AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 or Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.Chemical Identifiers
| eIF3A/eIF3S10 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| EIF3, p180 subunit; centrosomin homolog; cytoplasmic protein p167; eIF-3-theta; eIF3 p167; eIF3 p180; eIF3 p185; eukaryotic translation initiation factor 3 subunit 10; eukaryotic translation initiation factor 3 subunit A; eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD); eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD); eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa; eukaryotic translation initiation factor 3, subunit 10, 17 | |
| Between 860 and 910 | |
| Primary | |
| 4° C | |
| EIF3A |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P23116, Q14152 | |
| 13669, 8661 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Specifications
| eIF3A/eIF3S10 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P23116, Q14152 | |
| Eif3a | |
| Between 860 and 910 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| EIF3A | |
| EIF3, p180 subunit; centrosomin homolog; cytoplasmic protein p167; eIF-3-theta; eIF3 p167; eIF3 p180; eIF3 p185; eukaryotic translation initiation factor 3 subunit 10; eukaryotic translation initiation factor 3 subunit A; eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD); eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD); eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa; eukaryotic translation initiation factor 3, subunit 10, 17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13669, 8661 | |
| 4° C | |
| Liquid |