missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EIF2A Recombinant Rabbit Monoclonal Antibody (23GB1455)
GREENER_CHOICE

Catalog No. MA553162
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553162 100 μL
MA553163 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553162 Supplier Invitrogen™ Supplier No. MA553162

Rabbit Recombinant Monoclonal Antibody

EIF2A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.

Specifications

Antigen EIF2A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1455
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF2A
Gene Accession No. Q8BJW6, Q9BY44
Gene Alias 0910001O23Rik; 2410026C18Rik; 35kDa; 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; EIF2; EIF-2; EIF2A; eIF-2A; eIF2alpha; eIF-2alpha; eIF-2-alpha; EIF2S1; EIF2S1 protein; eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; eukaryotic translation initiation factor subunit 1-like protein; initiation factor 2 alpha; MST089; MSTP004; MSTP089
Gene Symbols EIF2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human eIF2A (450-510AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229317, 502531, 83939
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less