missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF1AX Recombinant Rabbit Monoclonal Antibody (23GB5290)
GREENER_CHOICE

Catalog No. MA552834
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552834 100 μL
MA552835 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552834 Supplier Invitrogen™ Supplier No. MA552834
Only null left

Rabbit Recombinant Monoclonal Antibody

eIF1AX Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.

Specifications

Antigen eIF1AX
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5290
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF1AX
Gene Accession No. P47813, Q8BMJ3
Gene Alias 1500010B24Rik; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; EIF1AP1; EIF1AX; Eif1ay; EIF4C; eIF-4C; eukaryotic translation initiation factor 1A X-linked; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A
Gene Symbols EIF1AX
Host Species Rabbit
Immunogen A synthesized peptide derived from human eIF1A (90-120AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1964, 302697, 66235
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less