missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic Acid Ethyl Ester Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015697
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015697 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015697 Supplier Merck Emd Millipore Supplier No. PHR17901G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 86227-47-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H34O2
Synonym cis-5,8,11,14,17-Eicosapentaenoic acid ethyl ester
Type Certified Reference Material
Quantity 1 g
Formula Weight 330.50
Grade Certified Reference Material