missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR4 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Early growth response protein 4 (EGR-4), also known as AT133, is a protein that in humans is encoded by the EGR4 gene. EGR-4 is a member of the early growth response (EGF) family of zinc finger transcription factors. Transcriptional regulator. Recognizes and binds to the DNA sequence 5'-GCGGGGGCG-3' (GSG). Activates the transcription of target genes whose products are required for mitogenesis and differentiation.
Specifications
Specifications
| Antigen | EGR4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | EGR4 |
| Gene Accession No. | Q05215 |
| Gene Alias | AT133; early growth response 4; early growth response protein 4; Early response protein NGFI-C; Egr4; EGR-4; Egr4l1; Nerve growth factor-induced protein C; NGF1-C; Ngfic; NGFI-C; PAT133; zinc-finger transcription factor NGFI-C (early response gene) |
| Gene Symbols | EGR4 |
| Show More |