missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGR3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5149783

Catalog No. PA5149783

Only null left
Explore available promotions

Description

Description

EGR3 Polyclonal Antibody for Western Blot, ICC/IF

This gene encodes a transcriptional regulator that belongs to the EGR family of C2H2-type zinc-finger proteins. It is an immediate-early growth response gene which is induced by mitogenic stimulation. The protein encoded by this gene participates in the transcriptional regulation of genes in controling biological rhythm. It may also play a role in a wide variety of processes including muscle development, lymphocyte development, endothelial cell growth and migration, and neuronal development. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Dec 2010].
Specifications

Specifications

EGR3
Polyclonal
Unconjugated
EGR3
early growth response 3; early growth response protein 3; EGR3; EGR-3; MGC138484; Pilot; zgc:92020; zinc finger protein pilot
Rabbit
Protein A
RUO
100516296, 1960, 25148
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P43301, Q06889
EGR3
Recombinant Early Growth Response Protein 3 (EGR3) (Cys98-Ala363).
200 μL
Primary
Human, Rat, Pig
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions