missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR1 Monoclonal Antibody (C15)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566020
Description
EGR1 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.Chemical Identifiers
| EGR1 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08046, P18146 | |
| 13653, 1958 | |
| Recombinant Early Growth Response Protein 1 (EGR1) (Gln282-Ser433). | |
| Antibody |
| C15 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human, Mouse | |
| EGR1 | |
| A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 | |
| EGR1 | |
| Primary | |
| Protein A/G |
Specifications
| EGR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08046, P18146 | |
| EGR1 | |
| Recombinant Early Growth Response Protein 1 (EGR1) (Gln282-Ser433). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C15 | |
| Unconjugated | |
| EGR1 | |
| A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 13653, 1958 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |