missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkey EGF ELISA Kit

ELISA

$930.00

Specifications

Accession Number NP
Assay Range 8.2 to 2000 pg/mL
Assay Sensitivity 8 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EPEGF
Encompass
View Documents Promotion Details
Invitrogen™
EPEGF
96 Tests
Pack for $930.00
Only null left
 
Description

Description

The Monkey Epidermal Growth Factor (Mk EGF) ELISA quantitates Rhesus Mk EGF in monkey serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Mk EGF. Principle of the method The Monkey EGF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
Specifications

Specifications

NP
8 pg/mL
HRP
GCPR
This ELISA pair antibody detects Rhesus Macaque EGF and human EGF.
Colorimetric Microplate Reader
EGF
<10
HRP
Inflammation
2°C to 8°C
EGF
4 hrs 45 mins
8.2 to 2000 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
698619
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
HOMG4, URG
1 hrs 20 mins
Videos
SDS
Documents

Documents