missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EGF ProQuantum Immunoassay Kit

Description
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
Specifications
Specifications
| Accession Number | P01133 |
| Assay Range | 0.0128-5000 pg/mL |
| Assay Sensitivity | <0.01 pg/mL |
| Content And Storage | Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5 to -30°C. |
| Conjugate | FAM |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| Show More |