missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Efficient-Pro™ Medium
Description
- A next generation CHO media used in the production of monoclonal antibodies
- Liquid medium offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
- Formulated as a chemically-defined, animal-origin free cell culture media used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
- Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
- Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively
- The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
- System is easier to use with less solubility issues than other CHO media and feed offerings
- Sustain cell viability for longer bioreactor runs & maximized titer
- Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Feed 1 compared to an industry-leader’s media and feed system
- Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
- Speed-up process development and streamline transfer to manufacturing scale
- Increase flexibility with a range of format & packaging configurations
- Decrease cell shock with simplified adaptation protocols
Specifications
Specifications
| Cell Line | CHO-DG44, CHO-S, CHO-K1, GS-CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Quantity | 1000 mL |
| Shipping Condition | Room temperature |
| Content And Storage | Store in the dark at 2–8°C. |
| Culture Type | Mammalian Cell Culture |
For research use only. Not for use in diagnostic procedures.