missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ Feed 1

Catalog No. A5208801
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5208801 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5208801 Supplier Gibco™ Supplier No. A5208801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco™ Efficient-Pro™ Feed 1 helps deliver superior product performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Feed 1 is chemically defined for maximum feeding flexibility and pairs best with feeds for CHO-K1, and CHO-DG44 cells.

  • A next generation CHO feed used in the production of monoclonal antibodies
  • Feed 1 offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Formulated as a chemically-defined, animal-origin free cell culture feed used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro™ Medium, Efficient-Pro™ Feed 1 has been formulated specifically for use in CHO-K1 cell lines
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Medium compared to an industry-leader’s media and feed system
  • Improve downstream outputs such as protein glycosylation, charge-variants, & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols

Specifications

Content And Storage Store in the dark at 2–8°C.

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Mammalian Cell Culture
Form Liquid
Quantity 1000 mL
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.