missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ DPM Feed Enhancer

Catalog No. A4000175502
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000175502 10 L
A4000175501 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4000175502 Supplier Gibco™ Supplier No. A4000175502
Only null left

Efficient-Pro DPM Feed Enhancer is a dry-format next-generation Chinese Hamster Ovary (CHO) feed enhancer used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Designed to work optimally with Efficient-Pro Medium and Efficient-Pro Feed 3, Efficient-Pro DPM Feed Enhancer has been formulated specifically for use in CHO-K1-GS cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use and scale with convenient formats available (liquid and DPM).

  • Increase ease-of-use with DPM dry format with simplified dissolution with minimal storage
  • Sustain cell viability for longer bioreactor runs and maximized titer
  • Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Ease of use with fast hydration time to save time on workflow

Efficient-Pro DPM Feed Enhancer has been formulated as a chemically defined, animal-origin-free cell culture feed supplement used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO-K1 cell line variants. Efficient-Pro DPM Feed Enhancer is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to support batch-to-batch consistency and production efficiency. It has been shown to improve productivity in a CHO-K1-GS cell line when paired with Efficient-Pro Feed 3 and Efficient-Pro Medium.

Specifications

Content And Storage Store in the dark at 2°C to 8°C.
Cell Line CHO-K1 GS, CHO-K1
Cell Type Mammalian Cell Culture
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Dehydrated, powder
Product Type CHO Media Supplement
Quantity 10 L
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.