missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ AGT™ Medium (+) Insulin

Catalog No. A4000458701
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
500 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000458701 1 L
A4000458702 10 L
A4000458703 100 L
A4000458704 500 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A4000458701 Supplier Gibco™ Supplier No. A4000458701

Efficient-Pro AGT Medium (+) Insulin is a dry-format next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Efficient-Pro AGT Medium (+) Insulin is a dry-format next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Designed to work optimally with the Efficient-Pro Feed system, Efficient-Pro AGT Medium (+) Insulin has been formulated specifically for use with insulin-dependent CHO cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality, and is easier to use and scale with convenient formats and multiple packaging configurations available.

Features of Efficient-Pro AGT Medium (+) Insulin include:

  • Benefit from the ease-of-use associated with Gibco Advanced Granulation technology (AGT) for auto-pH and simplified dissolution
  • Supports the growth and productivity of insulin-dependent CHO cell lines
  • Sustains cell viability for longer bioreactor runs and maximized titer
  • Helps speed up process development and streamline transfer to manufacturing scale
  • Increases flexibility with a range of format and packaging configurations

Efficient-Pro AGT Medium (+) Insulin is an animal-origin-free (AOF) cell culture medium based on the Efficient-Pro AGT Medium formulation with the addition of recombinant human insulin. It is designed for the bioproduction of mAbs and other therapeutic proteins in both batch and fed-batch bioprocessing workflows. It helps support the growth and productivity of insulin-dependent CHO cell lines by eliminating the need for manual insulin supplementation. This modification reduces handling steps, minimizes variability, and improves workflow efficiency. Designed to be paired with the Efficient-Pro Feed system, Efficient-Pro AGT Medium (+) Insulin provides the power to help achieve high titers, specific productivity, and the simplicity to start process development faster, facilitate easier adaptation, and transfer to manufacturing scale with ease.

Specifications

Cell Line Insulin-dependent CHO Cell Lines
Classification Chemically-defined, Animal Origin-free, Serum-free, cGMP
Form Powder (AGT)
Product Type CHO Media
Serum Level Serum-free
Product Line Next Gen CHO Media
Quantity 1 L
Shipping Condition Gel Packs
Green Features Sustainable packaging
Content And Storage Store in the dark at 2°C to 8°C.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.