missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ AGT™ Feed 1

Catalog No. A5209101
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5209101 1 L
A5209102 10 L
A5209103 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5209101 Supplier Gibco™ Supplier No. A5209101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco™ Efficient-Pro™ Feed 1 helps deliver superior performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Feed 1 is chemically defined for maximum feeding flexibility and pairs best with feeds for CHO-K1, and CHO-DG44 cells.

  • A next generation CHO feed used in the production of monoclonal antibodies
  • Feed 1 offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Formulated as a chemically-defined, animal-origin free cell culture feed used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors or insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro™ Medium, Efficient-Pro™ AGT™ Feed 1 has been formulated specifically for use in CHO-K1 cell lines
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Increase ease-of-use with Gibco’s™ trademarked, patented Advanced Granulation Technology™ (AGT™) for auto-pH and simplified dissolution
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Medium compared to an industry-leader’s media and feed system
  • Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols

Specifications

Content And Storage Store in the dark at 2–8°C.

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Mammalian Cell Culture
Form AGT
Quantity 1 L
Shipping Condition Room temperature

For research use only. Not for use in diagnostic procedures.