missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EFCAB3 Polyclonal Antibody

Description
Immunogen sequence: ADATTIKQHV KRATDTYNLG IALEHRKEML NLWQKIRGDL IGIDSRNE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
EFCAB3 gene ontology annotations related to this gene include calcium ion binding.
Specifications
Specifications
| Antigen | EFCAB3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EFCAB3 |
| Gene Accession No. | Q8N7B9 |
| Gene Alias | Efcab3; EF-hand calcium binding domain 3; EF-hand calcium-binding domain-containing protein 3; RGD1305423 |
| Gene Symbols | EFCAB3 |
| Show More |