missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EFCAB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569447 Supplier Invitrogen™ Supplier No. PA569447
Only null left

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

EFCAB14 gene ontology annotations related to this gene include calcium ion binding.

Specifications

Antigen EFCAB14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EFCAB14
Gene Accession No. Q8BGQ6
Gene Alias 4732418C07Rik; EFCAB14; EF-hand calcium binding domain 14; EF-hand calcium-binding domain-containing protein 14; Kiaa0494; RGD1310351; RP11-8J9.3
Gene Symbols EFCAB14
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of EFCAB14.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230648
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less