missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Efavirenz Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 391860-73-4 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C13H9 ClF3 N |
| Synonym | 6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 271.67 |
| Grade | Certified Reference Material |