missing translation for 'onlineSavingsMsg'
Learn More

Efavirenz Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002246
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002246 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002246 Supplier Merck Emd Millipore Supplier No. PHR206850MG
Only null left
Efavirenz USP Related Compound A is an impurity of the non-nucleoside HIV-1 reverse transcriptase inhibitor, efavirenz. Efavirenz is an antiretroviral medication used to treat and prevent Type-1 HIV.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 209414-27-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H11 ClF3 NO
Synonym (S)-1-(2-Amino-5-chlorophenyl)-1-(trifluoromethyl)-3-cyclopropyl-2-propyn-1-ol; (S)-2-(2-Amino-5-chlorophenyl)-4-cyclopropyl-1,1,1-trifluorobut-3-yn-2-ol
Type Certified Reference Material
Quantity 50 mg
Formula Weight 289.68
Grade Certified Reference Material