missing translation for 'onlineSavingsMsg'
Learn More

Efavirenz Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018407
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018407 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018407 Supplier Merck Emd Millipore Supplier No. PHR2072500MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 154598-52-4
Linear Formula C14H9 ClF3 NO2
MDL Number MFCD05662344
Synonym Efavirenz; (4 S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2 H-3,1-benzoxazin-2-one
Type Certified Reference Material
Quantity 500 mg
Formula Weight 315.67
Grade Certified Reference Material