missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Efaproxiral sodium, 98%

Catalog No. AC460100010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460100010 1 g
AC460102500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460100010 Supplier Thermo Scientific Chemicals Supplier No. 460100010
Only null left

CAS: 170787-99-2 | C20H22NNaO4 | 363.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170787-99-2
Molecular Formula C20H22NNaO4
Molecular Weight (g/mol) 363.38
InChI Key SWDPIHPGORBMFR-UHFFFAOYSA-M
Synonym efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13
PubChem CID 2725048
IUPAC Name sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate
SMILES CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+]

Specifications

Loss on Drying 13% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 363.38
Percent Purity 98%
Chemical Name or Material Efaproxiral sodium
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only