missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Titrant, 0.1 Molar M/10) Volumetric Solution, Standard, Macron Fine Chemicals™
macron
$558.78 - $558.78
Chemical Identifiers
| CAS | 139-33-3 |
|---|---|
| Molecular Formula | C10H14N2Na2O8 |
| Molecular Weight (g/mol) | 336.21 |
| MDL Number | MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 |
| InChI Key | ZGTMUACCHSMWAC-UHFFFAOYSA-L |
| Synonym | ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid |
| PubChem CID | 57339238 |
| ChEBI | CHEBI:64734 |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate |
| SMILES | [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Chemical Identifiers
| 139-33-3 | |
| 336.21 | |
| ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
| 57339238 | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate |
| C10H14N2Na2O8 | |
| MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
| ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
| CHEBI:64734 | |
| [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Specifications
| Colorless | |
| C10H14N2Na2O8 | |
| ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
| [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 336.21 | |
| CHEBI:64734 | |
| Liquid |
| 139-33-3 | |
| MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
| ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate | |
| 57339238 | |
| 1 L | |
| EDTA Titrant, 0.1 Molar (M/10) Volumetric Solution |