missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
Supplier: Ricca Chemical Company 27055
Specifications
| EDTA Titrant | |
| 6381-92-6,7732-18-5 | |
| 100.0,1.86 | |
| MFCD00150037,MFCD00003541 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| Cubitainer | |
| 20 L | |
| 0.0500Molar |
| Colorless | |
| 96.0,1.79 | |
| C10H18N2Na2O10 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| Laboratory | |
| 4 to 6 | |
| Liquid |
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |