missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate, (EDTA), 99-101%, Ultrapure

Catalog No. AAJ15700C4
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ15700C4 25 kg Plastic bottle
AAJ15700A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ15700C4 Supplier Thermo Fisher Scientific Supplier No. J15700C4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10378-23-1 ∣ C10H20N2Na4O10 ∣ 420.233 g/mol

Chemical Identifiers

CAS 10378-23-1
Molecular Formula C10H20N2Na4O10
Molecular Weight (g/mol) 420.233
MDL Number MFCD00150026
InChI Key DGWOWDJDIBPYOR-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate
PubChem CID 131843362
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na]

Specifications

Physical Form Crystalline Powder
Quantity 25 kg
Packaging Plastic bottle
Beilstein 3861753
Formula Weight 416.2
Percent Purity 99 to 101%
Grade Ungraded
Chemical Name or Material Ethylenediaminetetraacetic acid tetrasodium salt dihydrate
Hazard Category H302+H332-H318-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c
RTECSNumber AH4410000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only