missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ EDTA, OmniPur™, Calbiochem™,
$142.00 - $142.00
Chemical Identifiers
| CAS | 60-00-4 |
|---|---|
| Molecular Formula | C10H16N2O8 |
| Molecular Weight (g/mol) | 292.24 |
| MDL Number | MFCD00003541 |
| InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
| Synonym | edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol |
| PubChem CID | 6049 |
| ChEBI | CHEBI:42191 |
| IUPAC Name | 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Chemical Identifiers
| 60-00-4 | |
| 292.24 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 6049 | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| C10H16N2O8 | |
| MFCD00003541 | |
| edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
| CHEBI:42191 | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
| 60-00-4 | |
| White | |
| C10H16N2O8 | |
| 500 g | |
| Partially soluble in water | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
| 292.24 | |
| CHEBI:42191 | |
| ≥99.5% | |
| EDTA |
| 239.85°C | |
| 2.5 to 3.5 (aq. soln.) | |
| MFCD00003541 | |
| edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid | |
| 6049 | |
| 292.3 | |
| Solid | |
| Molecular biology grade |
Safety and Handling
Recommended Storage : Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.