Learn More
EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents
EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C22011003KG
Specifications
| EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate) | |
| >252°C | |
| White | |
| >252°C | |
| Poly Pail | |
| 3 kg | |
| Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate | |
| Pure |
| 6381-92-6 | |
| 252°C | |
| 4 to 6 | |
| 252°C | |
| C10H14N2O8Na2, 2H2O | |
| 1 | |
| 372.2369 | |
| Solid |