missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents
Specifications
Specifications
| Chemical Name or Material | EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate) |
| CAS | 6381-92-6 |
| Boiling Range | >252°C |
| Melting Point | 252°C |
| Color | White |
| pH | 4 to 6 |
| Boiling Point | >252°C |
| Melting Range | 252°C |
| Packaging | Poly Pail |
| Molecular Formula | C10H14N2O8Na2, 2H2O |
| Show More |