missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Disodium, 0.05 M Solution, Spectrum™ Chemical
$171.53 - $1780.09
Specifications
| Chemical Name or Material | EDTA Disodium |
|---|---|
| CAS | 6381-92-6,7732-18-5 |
| Concentration | 0.05 M ± 0.0005 M |
| Grade | Chemical Solution |
| Packaging | Cubitainer |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610333
|
Spectrum Chemical Mfg Cor
E112500MLPL |
500 mL | Cubitainer |
Each for $171.53
|
|
||||
|
18610332
|
Spectrum Chemical Mfg Cor
E1124LTPL |
4 L | Cubitainer |
Each for $338.47
|
|
||||
|
18610331
|
Spectrum Chemical Mfg Cor
E11220LTBL |
20 L | Cubitainer |
Each for $1,780.09
|
|
||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.