missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Certified, 0.1000M ±0.0005M (0.2N), LabChem™

Catalog No. LC139001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC139001 500 mL Poly Bottle
LC139002 1 L Poly Bottle
LC139004 4 L Poly Bottle
LC139006 10 L Cubitainer™
LC139005 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC139001 Supplier LabChem Supplier No. LC139001
Only null left

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 96.28, 3.72
Color Colorless
Density 1g/mL
Packaging Poly Bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Quantity 500 mL
Specific Gravity 1g/mL
Solubility Information Soluble in water
Formula Weight 372.23
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000M ±0.0005M (0.2N)
Physical Form Liquid
Chemical Name or Material EDTA
Show More Show Less
missing translation for 'healthHazard2' GHS H Statement
Solution is not hazardous.
missing translation for 'healthHazard3' GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature