missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, 0.5M (pH 8.0), Molecular Grade, Promega™
Chelator of divalent cations suitable for biochemistry and molecular biology applications.
$30.31 - $113.32
Chemical Identifiers
| Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
|---|---|
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
Description
- Available in 0.5M or 10mM concentrations
- 10mM concentration also provided as part of Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)
Chemical Identifiers
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| 962 | |
| CHEBI:15377 |
Specifications
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| CHEBI:15377 | |
| 100 mL |
| 962 | |
| Molecular Biology | |
| EDTA, 0.5M pH 8.0 |
Safety and Handling
Recommended Storage : 15 to 30°C