missing translation for 'onlineSavingsMsg'
Learn More

EDTA, 0.005 M, Reagents

Catalog No. 01335343
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335343 1 L
01335342 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335343 Supplier Labchem Inc Supplier No. CE2515001A

EDTA, 0.005 M, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6381-92-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt
IUPAC Name water
SMILES O

Specifications

pH 4 to 6
Boiling Point 100°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1.0009
Grade Reagent
Physical Form Liquid
Chemical Name or Material EDTA, 0.005 M