missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetate Disodium, Dihydrate, USP, 99-101%, Spectrum™ Chemical
ED150, 6381-92-6, C10H14N2Na2O8·2H2O
Supplier: Spectrum Chemical Mfg Cor ED150125GMCS6
Description
Spectrum™ Chemical Edetate Disodium, Dihydrate, USP is a chelating agent used to control heart rhythm disturbances caused by a heart medications. All Spectrum Chemical USP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 6381-92-6 | |
| 8.7-11.4% | |
| C10H18N2Na2O10 | |
| 6 x 125 g | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| USP |
| 100% | |
| Poly Bottle | |
| MFCD00150037,MFCD00003541 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 99 to 101% |
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |