missing translation for 'onlineSavingsMsg'
Learn More

Edetate Disodium, Dihydrate, USP, 99-101%, Spectrum™ Chemical

Catalog No. p-7157401
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602385 125 g Poly Bottle
18602387 500 g Poly Bottle
18602386 6 x 125 g Poly Bottle
18602389 2.5 kg Poly Pail
18602388 6 x 500 g Poly Bottle
18602390 4 x 2.5 kg Poly Pail
18602391 12 kg Poly Pail
18602392 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18602385 Supplier Spectrum Chemical Mfg Cor Supplier No. ED150125GM
Only null left

ED150, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Edetate Disodium, Dihydrate, USP is a chelating agent used to control heart rhythm disturbances caused by a heart medications. All Spectrum Chemical USP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 8.7-11.4%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99 to 101%
Grade USP