missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDAR Recombinant Mouse Monoclonal Antibody (EDAR12)
GREENER_CHOICE

Catalog No. MA551876
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551876 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551876 Supplier Invitrogen™ Supplier No. MA551876
Only null left

Mouse Recombinant Monoclonal Antibody

EDAR12 recognises and binds to CRD1 of the extracellular domain of mouse EDAR. EDAR3 cross-reacts with EDAR derived from human, dog, rat and chicken when EDAR is fused to the glycosylphosphatidylinositol anchor of TRAILR3. EDAR is the receptor for the TNF family ligand EDA1, which is a type II transmembrane protein possessing a collagen-like domain and a C-terminal TNF-homology domain. EDAR is important for the correct development of skin appendages including hair, teeth and eccrine sweat glands. LoF mutations in the Eda gene is known to cause XLHED (X-linked hypohidrotic ectodermal dysplasia), and results in abnormal development.

This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.

Specifications

Antigen EDAR
Applications ELISA, Functional Assay, Western Blot, Immunocytochemistry, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone EDAR12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Edar
Gene Accession No. Q9R187, Q9UNE0
Gene Alias Anhidrotic ectodysplasin receptor 1; DL; Downless; downless homolog; downless, mouse, homolog of; ECTD10A; ECTD10B; ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; ectodysplasin A receptor; ectodysplasin-A receptor; ED1R; ED3; ED5; EDA1R; EDA3; EDA-A1 receptor; EDA-A1R; Edar; HRM1; RGD1561714; Tumor necrosis factor receptor superfamily member EDAR
Gene Symbols Edar
Host Species Mouse
Immunogen EDAR12 was prepared by immunixzing female OVE1B mice (with the Edar gene deleted) subcutaneously with mouse EDAR-Fc and positive hybridoma clones were screened for binding mEDAR by ELISA.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10913, 13608, 365581, 428015, 481305
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less