missing translation for 'onlineSavingsMsg'
Learn More

Economical pH Test Strips

Line of pH test strips incorporates the most up-to-date technology.

Supplier:  Customer Channels Group (Ccg) PH30601B50

 View more versions of this product

Catalog No. S6113


Only null left

Description

Description

  • Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
  • Stabilized to provide the most accurate color determinations
  • Color charts printed on each pack for quick and easy identification
  • Each pack includes 50 strips

Specifications

Chemical Identifiers

Stabilized to provide user with most accurate color determination
50 Bag
pH Test Strip

Specifications

3 to 6
0.5
pH Test Strip
Stabilized to provide user with most accurate color determination
50 Bag
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 3 Years

SDS
Documents

Documents

Promotions

Promotions